MilliporeSigma
  • Non-imidazole histamine H3 ligands. Part III. New 4-n-propylpiperazines as non-imidazole histamine H3-antagonists.
Non-imidazole histamine H3 ligands. Part III. New 4-n-propylpiperazines as non-imidazole histamine H3-antagonists.
European journal of medicinal chemistry (2005-01-12)
Krzysztof Walczyński, Obbe P Zuiderveld, Henk Timmerman
ABSTRACT
In search for a new lead of non-imidazole histamine H3-receptor antagonists, a series of 1[(2-thiazolopyridine)-4-n-propyl]piperazines, the analogous 1-[(2-oxazolopyridine)-4-npropyl]piperazines, 1-[(2-benzothiazole)-4-n-propyl]piperazine and 1-[(2-benzooxazole)4-n-propyl]piperazine were prepared and in vitro tested as H3-receptor antagonists (the electrically evoked contraction of the guinea-pig jejunum). It appeared that by comparison of homologous pairs the thiazolo derivatives have slightly higher activity than their oxazolo analogues. The most potent compound of these series is the 1-(2-thiazolo[4,5-c]pyridine)-4-n-propylpiperazine (3c) with pA2 = 7.25 (its oxazole analogue (4g) showed pA2 = 6.9). The structure-activity relationships for compounds with various positions of the nitrogen in the benzene ring for the thiazoles compared with oxazoles are discussed.
Sigma-Aldrich
Sodium sulfide hydrate, ≥60%, scales
Pricing and availability is not currently available.