MilliporeSigma
  • Landscape of the regulatory elements for lysine 2-hydroxyisobutyrylation pathway.
Landscape of the regulatory elements for lysine 2-hydroxyisobutyrylation pathway.
Cell research (2017-12-02)
He Huang, Zhouqing Luo, Shankang Qi, Jing Huang, Peng Xu, Xiuxuan Wang, Li Gao, Fangyi Li, Jian Wang, Wenhui Zhao, Wei Gu, Zhucheng Chen, Lunzhi Dai, Junbiao Dai, Yingming Zhao
ABSTRACT
Short-chain fatty acids and their corresponding acyl-CoAs sit at the crossroads of metabolic pathways and play important roles in diverse cellular processes. They are also precursors for protein post-translational lysine acylation modifications. A noteworthy example is the newly identified lysine 2-hydroxyisobutyrylation (K
Sigma-Aldrich
MISSION® esiRNA, targeting human HDAC1
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution