MilliporeSigma
  • Ring-opening metathesis/oxy-cope rearrangement: a new strategy for the synthesis of bicyclic medium ring-containing compounds.
Ring-opening metathesis/oxy-cope rearrangement: a new strategy for the synthesis of bicyclic medium ring-containing compounds.
Journal of the American Chemical Society (2003-12-04)
Brian H White, Marc L Snapper
ABSTRACT
Ring-opening/ring-closing metathesis on cyclobutene-containing substrates with angular oxygen functionality provides a stereospecific introduction of 1,5-bis-dienes required for an anion-accelerated oxy-Cope rearrangement. The reaction sequence offers generally a stereocontrolled preparation of a variety of medium ring-containing bicyclic ring systems, while rearrangement to the bicyclo[7,3,0]dodecane (9-5) system leads to a mixture of olefin isomers.
Product Description
Hoveyda-Grubbs Catalyst® M720 ChemBeads
Pricing and availability is not currently available.
Sigma-Aldrich
Hoveyda-Grubbs Catalyst® M720, Umicore, 97%
Pricing and availability is not currently available.