MilliporeSigma
  • STIM1 promotes migration, phagosomal maturation and antigen cross-presentation in dendritic cells.
STIM1 promotes migration, phagosomal maturation and antigen cross-presentation in dendritic cells.
Nature communications (2017-11-28)
Paula Nunes-Hasler, Sophia Maschalidi, Carla Lippens, Cyril Castelbou, Samuel Bouvet, Daniele Guido, Flavien Bermont, Esen Y Bassoy, Nicolas Page, Doron Merkler, Stéphanie Hugues, Denis Martinvalet, Bénédicte Manoury, Nicolas Demaurex
ABSTRACT
Antigen cross-presentation by dendritic cells (DC) stimulates cytotoxic T cell activation to promote immunity to intracellular pathogens, viruses and cancer. Phagocytosed antigens generate potent T cell responses, but the signalling and trafficking pathways regulating their cross-presentation are unclear. Here, we show that ablation of the store-operated-Ca2+-entry regulator STIM1 in mouse myeloid cells impairs cross-presentation and DC migration in vivo and in vitro. Stim1 ablation reduces Ca2+ signals, cross-presentation, and chemotaxis in mouse bone-marrow-derived DCs without altering cell differentiation, maturation or phagocytic capacity. Phagosomal pH homoeostasis and ROS production are unaffected by STIM1 deficiency, but phagosomal proteolysis and leucyl aminopeptidase activity, IRAP recruitment, as well as fusion of phagosomes with endosomes and lysosomes are all impaired. These data suggest that STIM1-dependent Ca2+ signalling promotes the delivery of endolysosomal enzymes to phagosomes to enable efficient cross-presentation.
Sigma-Aldrich
Anti-ORAI1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-STIM1 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-IRAP Antibody, Clone 3E1, clone 3E1, from mouse