MilliporeSigma
  • A Ribosomopathy Reveals Decoding Defective Ribosomes Driving Human Dysmorphism.
A Ribosomopathy Reveals Decoding Defective Ribosomes Driving Human Dysmorphism.
American journal of human genetics (2017-03-05)
Nahuel A Paolini, Martin Attwood, Samuel B Sondalle, Carolina Marques Dos Santos Vieira, Anita M van Adrichem, Franca M di Summa, Marie-Françoise O'Donohue, Pierre-Emmanuel Gleizes, Swaksha Rachuri, Joseph W Briggs, Roman Fischer, Peter J Ratcliffe, Marcin W Wlodarski, Riekelt H Houtkooper, Marieke von Lindern, Taco W Kuijpers, Jonathan D Dinman, Susan J Baserga, Matthew E Cockman, Alyson W MacInnes
ABSTRACT
Ribosomal protein (RP) gene mutations, mostly associated with inherited or acquired bone marrow failure, are believed to drive disease by slowing the rate of protein synthesis. Here de novo missense mutations in the RPS23 gene, which codes for uS12, are reported in two unrelated individuals with microcephaly, hearing loss, and overlapping dysmorphic features. One individual additionally presents with intellectual disability and autism spectrum disorder. The amino acid substitutions lie in two highly conserved loop regions of uS12 with known roles in maintaining the accuracy of mRNA codon translation. Primary cells revealed one substitution severely impaired OGFOD1-dependent hydroxylation of a neighboring proline residue resulting in 40S ribosomal subunits that were blocked from polysome formation. The other disrupted a predicted pi-pi stacking interaction between two phenylalanine residues leading to a destabilized uS12 that was poorly tolerated in 40S subunit biogenesis. Despite no evidence of a reduction in the rate of mRNA translation, these uS12 variants impaired the accuracy of mRNA translation and rendered cells highly sensitive to oxidative stress. These discoveries describe a ribosomopathy linked to uS12 and reveal mechanistic distinctions between RP gene mutations driving hematopoietic disease and those resulting in developmental disorders.
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Glucose-6-Phosphate Dehydrogenase (G-6-PDH) antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
Sigma-Aldrich
Anti-OGFOD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture