MilliporeSigma
Mena
Scientific reports (2016-11-09)
Maxwell D Weidmann, Chinmay R Surve, Robert J Eddy, Xiaoming Chen, Frank B Gertler, Ved P Sharma, John S Condeelis
ABSTRACT
Invadopodia, actin-based protrusions of invasive carcinoma cells that focally activate extracellular matrix-degrading proteases, are essential for the migration and intravasation of tumor cells during dissemination from the primary tumor. We have previously shown that cortactin phosphorylation at tyrosine residues, in particular tyrosine 421, promotes actin polymerization at newly-forming invadopodia, promoting their maturation to matrix-degrading structures. However, the mechanism by which cells regulate the cortactin tyrosine phosphorylation-dephosphorylation cycle at invadopodia is unknown. Mena, an actin barbed-end capping protein antagonist, is expressed as various splice-isoforms. The Mena
Sigma-Aldrich
Anti-phospho-Cortactin (pTyr421) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
PTP1B Inhibitor, PTP1B Inhibitor, CAS 765317-72-4, is a cell-permeable, selective, reversible and non-competitive allosteric inhibitor of PTP1B (IC₅₀ = 4 µM and 8 µM for PTP1B403 and PTP1B298, respectively).