MilliporeSigma
  • NNT is a key regulator of adrenal redox homeostasis and steroidogenesis in male mice.
NNT is a key regulator of adrenal redox homeostasis and steroidogenesis in male mice.
The Journal of endocrinology (2017-10-20)
E Meimaridou, M Goldsworthy, V Chortis, E Fragouli, P A Foster, W Arlt, R Cox, L A Metherell
ABSTRACT
Nicotinamide nucleotide transhydrogenase, NNT, is a ubiquitous protein of the inner mitochondrial membrane with a key role in mitochondrial redox balance. NNT produces high concentrations of NADPH for detoxification of reactive oxygen species by glutathione and thioredoxin pathways. In humans, NNT dysfunction leads to an adrenal-specific disorder, glucocorticoid deficiency. Certain substrains of C57BL/6 mice contain a spontaneously occurring inactivating
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-NNT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CYP21A2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution