MilliporeSigma
SEC23B is required for pancreatic acinar cell function in adult mice.
Molecular biology of the cell (2017-05-26)
Rami Khoriaty, Nancy Vogel, Mark J Hoenerhoff, M Dolors Sans, Guojing Zhu, Lesley Everett, Bradley Nelson, Haritha Durairaj, Brooke McKnight, Bin Zhang, Stephen A Ernst, David Ginsburg, John A Williams
ABSTRACT
Mice with germline absence of SEC23B die perinatally, exhibiting massive pancreatic degeneration. We generated mice with tamoxifen-inducible, pancreatic acinar cell-specific
Sigma-Aldrich
Anti-α-Amylase antibody produced in rabbit, fractionated antiserum, lyophilized powder