MilliporeSigma
Txndc9 Is Required for Meiotic Maturation of Mouse Oocytes.
BioMed research international (2017-06-20)
Fanhua Ma, Liming Hou, Liguo Yang
ABSTRACT
Txndc9 (thioredoxin domain containing protein 9) has been shown to be involved in mammalian mitosis; however, its function in mammalian oocyte meiosis remains unclear. In this study, we initially found that Txndc9 is expressed during meiotic maturation of mouse oocytes and higher expression of Txndc9 mRNA and protein occurred in germinal vesicle (GV) stage. By using confocal scanning, we observed that Txndc9 localized at both nucleus and cytoplasm, especially at spindle microtubules. Specific depletion of Txndc9 by siRNA in mouse oocyte resulted in decreasing the rate of first polar body extrusion and increasing abnormal spindle assemble. Moreover, knockdown of Txndc9 in germinal vesicle (GV) stage oocytes led to higher level of reactive oxygen species (ROS) and lower level of antioxidant glutathione (GSH) as compared with control oocytes, which indicated that Txndc9 may be involved in mediating the redox balance. In summary, our results demonstrated that Txndc9 is crucial for mouse oocyte maturation by regulating spindle assembly, polar body extrusion, and redox status.
Sigma-Aldrich
Anti-TXNDC9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution