MilliporeSigma
  • Dynamic Arc SUMOylation and Selective Interaction with F-Actin-Binding Protein Drebrin A in LTP Consolidation
Dynamic Arc SUMOylation and Selective Interaction with F-Actin-Binding Protein Drebrin A in LTP Consolidation
Frontiers in synaptic neuroscience (2017-05-30)
Rajeevkumar R Nair, Sudarshan Patil, Adrian Tiron, Tambudzai Kanhema, Debabrata Panja, Lars Schiro, Kamil Parobczak, Grzegorz Wilczynski, Clive R Bramham
ABSTRACT
Activity-regulatedcytoskeleton-associated protein (Arc) protein is implicated as a master regulator of long-term forms of synaptic plasticity and memory formation, but the mechanisms controlling Arc protein function are little known. Post-translation modification by small ubiquitin-like modifier (SUMO) proteins has emerged as a major mechanism for regulating protein-protein interactions and function. We first show in cell lines that ectopically expressed Arc undergoes mono-SUMOylation. The covalent addition of a single SUMO1 protein was confirmed by
Sigma-Aldrich
Anti-β-Actin–FITC antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture