MilliporeSigma
  • Integrins Modulate T Cell Receptor Signaling by Constraining Actin Flow at the Immunological Synapse.
Integrins Modulate T Cell Receptor Signaling by Constraining Actin Flow at the Immunological Synapse.
Frontiers in immunology (2018-02-07)
Katarzyna I Jankowska, Edward K Williamson, Nathan H Roy, Daniel Blumenthal, Vidhi Chandra, Tobias Baumgart, Janis K Burkhardt
ABSTRACT
Full T cell activation requires coordination of signals from multiple receptor-ligand pairs that interact in parallel at a specialized cell-cell contact site termed the immunological synapse (IS). Signaling at the IS is intimately associated with actin dynamics; T cell receptor (TCR) engagement induces centripetal flow of the T cell actin network, which in turn enhances the function of ligand-bound integrins by promoting conformational change. Here, we have investigated the effects of integrin engagement on actin flow, and on associated signaling events downstream of the TCR. We show that integrin engagement significantly decelerates centripetal flow of the actin network. In primary CD4
Sigma-Aldrich
Anti-ACTN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution