MilliporeSigma
  • LRG1 promotes proliferation and inhibits apoptosis in colorectal cancer cells via RUNX1 activation.
LRG1 promotes proliferation and inhibits apoptosis in colorectal cancer cells via RUNX1 activation.
PloS one (2017-04-05)
Ying Zhou, Xintian Zhang, Jingjing Zhang, Jingyuan Fang, Zhizheng Ge, Xiaobo Li
ABSTRACT
Leucine-rich-alpha-2-glycoprotein 1 (LRG1) has been shown to be involved in various human malignancies. Whether it plays a role in colorectal cancer (CRC) development remains unclear. Here, we investigated whether and through what mechanism LRG1 functions in human CRC cells. The plasma level of LRG1 was significantly increased in CRC patients, but it was remarkably decreased in patients with resected colorectal cancers. Meanwhile, both mRNA and protein levels of LRG1 were remarkable overexpressed in CRC tissues than normal tissues. The knockdown of LRG1 significantly inhibited cell proliferation, induced cell cycle arrest at the G0/G1 phase, and promoted apoptosis in SW480 and HCT116 cells in vitro. In addition, LRG1 silencing led to the downregulation of the levels of key cell cycle factors, such as cyclin D1, B, and E and anti-apoptotic B-cell lymphoma-2(Bcl-2). However, it up-regulated the expression of pro-apoptotic Bax and cleaved caspase-3. Furthermore, RUNX1 could be induced by LRG1 in a concentration-dependent manner, while the knockdown of RUNX1 blocked the promotion of the proliferation and inhibition of apoptosis induced by LRG1. Collectively, these findings indicate that LRG1 plays a crucial role in the proliferation and apoptosis of CRC by regulating RUNX1 expression. Thus, LRG1 may be a potential detection biomarker as well as a marker for monitoring recurrence and therapeutic target for CRC.
Sigma-Aldrich
Anti-LRG1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution