MilliporeSigma
  • Meru couples planar cell polarity with apical-basal polarity during asymmetric cell division.
Meru couples planar cell polarity with apical-basal polarity during asymmetric cell division.
eLife (2017-07-01)
Jennifer J Banerjee, Birgit L Aerne, Maxine V Holder, Simon Hauri, Matthias Gstaiger, Nicolas Tapon
ABSTRACT
Polarity is a shared feature of most cells. In epithelia, apical-basal polarity often coexists, and sometimes intersects with planar cell polarity (PCP), which orients cells in the epithelial plane. From a limited set of core building blocks (e.g. the Par complexes for apical-basal polarity and the Frizzled/Dishevelled complex for PCP), a diverse array of polarized cells and tissues are generated. This suggests the existence of little-studied tissue-specific factors that rewire the core polarity modules to the appropriate conformation. In
Sigma-Aldrich
Monoclonal ANTI-FLAG® antibody produced in rabbit, clone SIG1-25, ascites fluid