MilliporeSigma
  • Temporal progression of PARP activity in the Prph2 mutant rd2 mouse: Neuroprotective effects of the PARP inhibitor PJ34.
Temporal progression of PARP activity in the Prph2 mutant rd2 mouse: Neuroprotective effects of the PARP inhibitor PJ34.
PloS one (2017-07-21)
Ayse Sahaboglu, Alaa Sharif, Lili Feng, Enver Secer, Eberhart Zrenner, François Paquet-Durand
ABSTRACT
Peripherin (peripherin/rds) is a membrane-associated protein that plays a critical role in the morphogenesis of rod and cone photoreceptor outer segments. Mutations in the corresponding PRPH2 gene cause different types of retinal dystrophies characterized by a loss of photoreceptors. Over activation of poly-ADP-ribose polymerase (PARP) was previously shown to be involved in different animal models for hereditary retinal dystrophies. This includes the rd2 mouse, which suffers from a human homologous mutation in the PRPH2 gene. In the present study, we show that increased retinal PARP activity and poly-ADP-ribosylation of proteins occurs before the peak of rd2 photoreceptor degeneration. Inhibition of PARP activity with the well-characterized PARP inhibitor PJ34 decreased the levels of poly-ADP-ribosylation and photoreceptor cell death. These results suggest a causal involvement of PARP in photoreceptor degeneration caused by peripherin mutations and highlight the possibility to use PARP inhibition for the mutation-independent treatment of hereditary retinal dystrophies.
Sigma-Aldrich
Anti-Rhodopsin Antibody, clone RET-P1, clone RET-P1, Chemicon®, from mouse