MilliporeSigma
  • Distinct molecular mechanisms of acquired resistance to temozolomide in glioblastoma cells.
Distinct molecular mechanisms of acquired resistance to temozolomide in glioblastoma cells.
Journal of neurochemistry (2012-05-09)
Caroline Happold, Patrick Roth, Wolfgang Wick, Natalie Schmidt, Ana-Maria Florea, Manuela Silginer, Guido Reifenberger, Michael Weller
ABSTRACT
Temozolomide (TMZ) is an alkylating chemotherapeutic agent that prolongs the survival of patients with glioblastoma. Clinical benefit is more prominent in patients with methylation of the O(6) -methyl-guanine DNA methyltransferase (MGMT) promoter. However, all patients eventually suffer from tumor progression because their tumors become resistant to TMZ. Here, we modeled acquired TMZ resistance in glioma cells in vitro to identify underlying molecular mechanisms. To this end, the glioma cell lines LNT-229, LN-308, and LN-18 were exposed repetitively to increasing concentrations of TMZ to induce a stable resistant phenotype (R) defined by clonogenic survival assays. The molecular mechanisms mediating acquired resistance were assessed by immunoblot, PCR, and flow cytometry. Rescue experiments were performed with siRNA-mediated candidate gene silencing. We found in LN-18 cells constitutively expressing MGMT a strong up-regulation of MGMT levels in TMZ-resistant cells. TMZ resistance in the MGMT-negative cell lines LNT-229 and LN-308 was not associated with de novo expression of MGMT. Instead, we found a down-regulation of several DNA mismatch-repair proteins in resistant LNT-229 cells. A TMZ-resistant phenotype was also achieved by silencing selected DNA mismatch repair proteins in parental LNT-229 cells. No obvious mechanism of resistance was identified in the third cell line, LN-308, except for reduced methylation of LINE-1 repetitive elements. In conclusion, we demonstrate that different molecular mechanisms may contribute to the development of acquired TMZ resistance in glioma cells, indicating the need to develop distinct strategies to overcome resistance.
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
ChIPAb+ Histone H3 (CT) - ChIP Validated Antibody and Primer Set, rabbit monoclonal, culture supernatant, from rabbit
Sigma-Aldrich
ChIPAb+ Trimethyl-Histone H3 (Lys9) - ChIP Validated Antibody and Primer Set, from rabbit