MilliporeSigma
  • Micro-adhesion rings surrounding TCR microclusters are essential for T cell activation.
Micro-adhesion rings surrounding TCR microclusters are essential for T cell activation.
The Journal of experimental medicine (2016-06-30)
Akiko Hashimoto-Tane, Machie Sakuma, Hiroshi Ike, Tadashi Yokosuka, Yayoi Kimura, Osamu Ohara, Takashi Saito
ABSTRACT
The immunological synapse (IS) formed at the interface between T cells and antigen-presenting cells represents a hallmark of initiation of acquired immunity. T cell activation is initiated at T cell receptor (TCR) microclusters (MCs), in which TCRs and signaling molecules assemble at the interface before IS formation. We found that each TCR-MC was transiently bordered by a ring structure made of integrin and focal adhesion molecules in the early phase of activation, which is similar in structure to the IS in microscale. The micro-adhesion ring is composed of LFA-1, focal adhesion molecules paxillin and Pyk2, and myosin II (MyoII) and is supported by F-actin core and MyoII activity through LFA-1 outside-in signals. The formation of the micro-adhesion ring was transient but especially sustained upon weak TCR stimulation to recruit linker for activation of T cells (LAT) and SLP76. Perturbation of the micro-adhesion ring induced impairment of TCR-MC development and resulted in impaired cellular signaling and cell functions. Thus, the synapse-like structure composed of the core TCR-MC and surrounding micro-adhesion ring is a critical structure for initial T cell activation through integrin outside-in signals.
Sigma-Aldrich
Anti-Vinculin−FITC antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
Pricing and availability is not currently available.