MilliporeSigma
  • RNASeq analysis of differentiated keratinocytes reveals a massive response to late events during human papillomavirus type 16 infection, including loss of epithelial barrier function.
RNASeq analysis of differentiated keratinocytes reveals a massive response to late events during human papillomavirus type 16 infection, including loss of epithelial barrier function.
Journal of virology (2017-10-13)
T Klymenko, Q Gu, I Herbert, A Stevenson, V Iliev, G Watkins, C Pollock, R Bhatia, K Cuschieri, P Herzyk, D Gatherer, S V Graham
ABSTRACT
The human papillomavirus (HPV) replication cycle is tightly linked to epithelial cell differentiation. To examine HPV-associated changes in the keratinocyte transcriptome, RNAs isolated from undifferentiated and differentiated cell populations of normal, spontaneously immortalised, keratinocytes (NIKS), and NIKS stably transfected with HPV16 episomal genomes (NIKS16), were compared using RNASeq. HPV16 infection altered expression of 2862 cellular genes. Next, to elucidate the role of keratinocyte gene expression in late events during the viral life cycle, RNASeq was carried out on triplicate differentiated populations of NIKS (uninfected) and NIKS16 (infected). Of the top 966 genes altered (>log
Sigma-Aldrich
Monoclonal Anti-Involucrin antibody produced in mouse, clone SY5, ascites fluid