MilliporeSigma
Live imaging looks deeper.
eLife (2017-09-05)
Tanner C Fadero, Paul S Maddox
ABSTRACT
Iodixanol provides an easy and affordable solution to a problem that has limited resolution and brightness when imaging living samples.
Sigma-Aldrich
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation