MilliporeSigma
  • The interaction between fibrinogen and zymogen FXIII-A2B2 is mediated by fibrinogen residues γ390-396 and the FXIII-B subunits.
The interaction between fibrinogen and zymogen FXIII-A2B2 is mediated by fibrinogen residues γ390-396 and the FXIII-B subunits.
Blood (2016-08-27)
James R Byrnes, Clare Wilson, Anthony M Boutelle, Chase B Brandner, Matthew J Flick, Helen Philippou, Alisa S Wolberg
ABSTRACT
Coagulation transglutaminase factor XIII (FXIII) exists in circulation as heterotetrameric proenzyme FXIII-A
Sigma-Aldrich
Anti-F13B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution