MilliporeSigma
MK2 Phosphorylates RIPK1 to Prevent TNF-Induced Cell Death.
Molecular cell (2017-05-17)
Isabel Jaco, Alessandro Annibaldi, Najoua Lalaoui, Rebecca Wilson, Tencho Tenev, Lucie Laurien, Chun Kim, Kunzah Jamal, Sidonie Wicky John, Gianmaria Liccardi, Diep Chau, James M Murphy, Gabriela Brumatti, Rebecca Feltham, Manolis Pasparakis, John Silke, Pascal Meier
ABSTRACT
TNF is an inflammatory cytokine that upon binding to its receptor, TNFR1, can drive cytokine production, cell survival, or cell death. TNFR1 stimulation causes activation of NF-κB, p38α, and its downstream effector kinase MK2, thereby promoting transcription, mRNA stabilization, and translation of target genes. Here we show that TNF-induced activation of MK2 results in global RIPK1 phosphorylation. MK2 directly phosphorylates RIPK1 at residue S321, which inhibits its ability to bind FADD/caspase-8 and induce RIPK1-kinase-dependent apoptosis and necroptosis. Consistently, a phospho-mimetic S321D RIPK1 mutation limits TNF-induced death. Mechanistically, we find that phosphorylation of S321 inhibits RIPK1 kinase activation. We further show that cytosolic RIPK1 contributes to complex-II-mediated cell death, independent of its recruitment to complex-I, suggesting that complex-II originates from both RIPK1 in complex-I and cytosolic RIPK1. Thus, MK2-mediated phosphorylation of RIPK1 serves as a checkpoint within the TNF signaling pathway that integrates cell survival and cytokine production.
Sigma-Aldrich
GSK′963, ≥98% (HPLC)
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Monoclonal Anti-α1-Acid Glycoprotein antibody produced in mouse, clone AGP-47, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)