MilliporeSigma
  • Requirement of Gαq/Gα11 Signaling in the Preservation of Mouse Intestinal Epithelial Homeostasis.
Requirement of Gαq/Gα11 Signaling in the Preservation of Mouse Intestinal Epithelial Homeostasis.
Cellular and molecular gastroenterology and hepatology (2017-02-09)
Noboru Watanabe, Hirosato Mashima, Kouichi Miura, Takashi Goto, Makoto Yoshida, Akiteru Goto, Hirohide Ohnishi
ABSTRACT
Proliferation, differentiation, and morphogenesis of the intestinal epithelium are tightly regulated by a number of molecular pathways. Coordinated action of intestine is achieved by gastrointestinal hormones, most of which exert these actions through G-protein-coupled receptors. We herein investigated the role of Gαq/11-mediated signaling in intestinal homeostasis. Intestinal tissues from control (Gnaq flox/flox Gna11 +/+ ), Int-Gq knock-out (KO) (VilCre +/- Gnaq flox/flox Gna11 +/+ ), G11 KO (Gnaq flox/flox Gna11 -/- ), and Int-Gq/G11 double knock-out (DKO) (VilCre +/- Gnaq flox/flox Gna11 -/- ) mice were examined by microscopy, transmission electron microscopy, and immunohistochemistry. The effect of Gαq/11-mediated signaling was studied in the cell lineage, proliferation, and apoptosis. Dextran sodium sulfate (DSS) colitis was induced to study the role of Gαq/11 in colon. Paneth cells were enlarged, increased in number, and mislocalized in Int-Gq/G11 DKO small intestine. Paneth cells also reacted with PAS and Muc2 antibody, indicating an intermediate character of Paneth and goblet cells. The nuclear β-catenin, T-cell factor 1, and Sox9 expression were reduced severely in the crypt base of Int-Gq/G11 DKO intestine. Proliferation was activated in the crypt base and apoptosis was enhanced along the crypt. Int-Gq/G11 DKO mice were susceptible to DSS colitis. Proliferation was inhibited in the crypt of unaffected and regenerative areas. Cystic crypts, periodic acid-Schiff-positive cells, and Muc2-positive cells were unusually observed in the ulcerative region. The Gαq/11-mediated pathway plays a pivotal role in the preservation of intestinal homeostasis, especially in Paneth cell maturation and positioning. Wnt/β-catenin signaling was reduced significantly in the crypt base in Gαq/G11-deficient mice, resulting in the defective maturation of Paneth cells, induction of differentiation toward goblet cells, and susceptibility to DSS colitis.
Sigma-Aldrich
Anti-HES-5 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-BrdU antibody produced in mouse, clone BU-33, ascites fluid, Immunohistology Grade
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit