- Identification of proteins with different abundance associated with cell migration and proliferation in leiomyoma interstitial fluid by proteomics.
Identification of proteins with different abundance associated with cell migration and proliferation in leiomyoma interstitial fluid by proteomics.
Oncology letters (2017-05-20)
Blendi Ura, Federica Scrimin, Cinzia Franchin, Giorgio Arrigoni, Danilo Licastro, Lorenzo Monasta, Giuseppe Ricci
PMID28521489
ABSTRACT
Uterine leiomyoma is the most common female reproductive tract benign tumor. Little is known about protein composition and changes in the leiomyoma interstitial fluid (IF). The present study focused on changes in protein abundance in the IF of leiomyoma. Leiomyoma IFs and adjacent myometrial IFs were obtained and analyzed by two-dimensional electrophoresis (2-DE) coupled with mass spectrometry and western blotting for 2-DE data validation. A total of 25 unique proteins were observed to change significantly (P<0.05). Of these proteins with different abundance, 22 had not been previously identified in leiomyoma IF.
Sigma-Aldrich
Anti-PRDX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-LUM antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-ANXA2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution