MilliporeSigma
  • Comprehensive Spatial Analysis of the Borrelia burgdorferi Lipoproteome Reveals a Compartmentalization Bias toward the Bacterial Surface.
Comprehensive Spatial Analysis of the Borrelia burgdorferi Lipoproteome Reveals a Compartmentalization Bias toward the Bacterial Surface.
Journal of bacteriology (2017-01-11)
Alexander S Dowdell, Maxwell D Murphy, Christina Azodi, Selene K Swanson, Laurence Florens, Shiyong Chen, Wolfram R Zückert
ABSTRACT
The Lyme disease spirochete
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution