MilliporeSigma
  • Iron-catalysed fluoroaromatic coupling reactions under catalytic modulation with 1,2-bis(diphenylphosphino)benzene.
Iron-catalysed fluoroaromatic coupling reactions under catalytic modulation with 1,2-bis(diphenylphosphino)benzene.
Chemical communications (Cambridge, England) (2009-02-26)
Takuji Hatakeyama, Yoshiyuki Kondo, Yu-Ichi Fujiwara, Hikaru Takaya, Shingo Ito, Eiichi Nakamura, Masaharu Nakamura
ABSTRACT
A catalytic amount of 1,2-bis(diphenylphosphino)benzene (DPPBz) achieves selective cleavage of sp(3)-carbon-halogen bond in the iron-catalysed cross-coupling between polyfluorinated arylzinc reagents and alkyl halides, which was unachievable with a stoichiometric modifier such as TMEDA; the selective iron-catalysed fluoroaromatic coupling provides easy and practical access to polyfluorinated aromatic compounds.