MilliporeSigma
DDEFL1 correlated with Rho GTPases activity in breast cancer.
Oncotarget (2018-01-20)
Xiaoyun Mao, Chuifeng Fan, Xinmiao Yu, Bo Chen, Feng Jin
ABSTRACT
DDEFL1 is related to maintaining a limiting amount of ARF6 in GTP-loaded form by accelerating its GTP hydrolysis activity, which has been implicated in hepatocellular cancer pathogenesis and lung cancer development. We investigated DDEFL1 expression in breast cancer and paired normal breast tissues by immunohistochemistry and found that DDEFL1 expression was significantly associated with tumor size, lymph node metastasis, high content of elastosis and TNM stage but not with menopausal status or age. We detected the mRNA and protein expression of DDEFL1 in breast cancer cell lines by Western blotting and quantitative real-time PCR (qRT-PCR). DDEFL1 was obvious in MDA-MB-435s and MDA-MB-231 but very weak in ZR-75-1. Further experiments were conducted to evaluate the effect of DDEFL1 small interfering RNA (siRNA) transfection on the biological behavior of MDA-MB-231. After transfection, the effects of DDEFL1 inhibition on expression of mRNA and protein were also analyzed by Western blotting and qRT-PCR. Increased apoptosis and invasive ability, decreased cellular proliferation was found in MDA-MB-231 with successful DDEFL1 siRNA transient transfection (
Sigma-Aldrich
MISSION® esiRNA, targeting human ASAP3