MilliporeSigma
  • Fluorescence quenching-based signal amplification on immunochromatography test strips for dual-mode sensing of two biomarkers of breast cancer.
Fluorescence quenching-based signal amplification on immunochromatography test strips for dual-mode sensing of two biomarkers of breast cancer.
Nanoscale (2017-11-23)
Bo Zhang, Wenjuan Ma, Fuxin Li, Weichen Gao, Qian Zhao, Weipan Peng, Jiafang Piao, Xiaoli Wu, Hanjie Wang, Xiaoqun Gong, Jin Chang
ABSTRACT
Recently, immunochromatography test strips (ICTS) have been fully developed for point-of-care testing (POCT). However, the intrinsic limitations including non-quantitative detection of colloidal gold ICTS and low sensitivity of fluorescence ICTS (FICTS) significantly restrict their further application in clinical diagnosis. Taking advantages of rapid colorimetric qualitative detection and fluorescence quantitation, we designed a kind of sensitive and dual-mode magnetic FICTS (mFICTS) based on PLGA@Fe3O4 super-paramagnetic nanosphere (SPMN) probes quenching multiplex fluorescer on the test line through sandwich immunoreactions. Owing to the large number of Fe
Sigma-Aldrich
1,2-Dodecanediol, 90%