MilliporeSigma
  • Development of colloidally stable carbazole-based fluorescent nanoaggregates.
Development of colloidally stable carbazole-based fluorescent nanoaggregates.
Journal of photochemistry and photobiology. A, Chemistry (2018-02-13)
Denis Svechkarev, Alexander Kyrychenko, William M Payne, Aaron M Mohs
ABSTRACT
Fluorescent nanomaterials require high colloidal stability for effective use in imaging and sensing applications. We herein report the synthesis of carbazole-based organic fluorescent nanoaggregates, and demonstrate the superior colloidal stability of alkyl-substituted dye aggregates over their non-alkylated analogs. The role of alkyl chains in self-assembly and stability of such nanoaggregates is discussed based on both experimental and molecular dynamics simulation data, and spectral characteristics of the precursor dyes and their aggregates are described. The obtained results provide new insights on development of colloidally stable organic fluorescent nanomaterials with low polydispersity.
Sigma-Aldrich
DCC, 99%
Sigma-Aldrich
1,4-Dibromobenzene, 98%
Sigma-Aldrich
3,6-Dibromocarbazole, 97%
Sigma-Aldrich
5-(Dihydroxyboryl)-2-thiophenecarboxylic acid, AldrichCPR