MilliporeSigma
  • Proteinase activated receptor-2 counterbalances the vascular effects of endothelin-1 in fibrotic tight-skin mice.
Proteinase activated receptor-2 counterbalances the vascular effects of endothelin-1 in fibrotic tight-skin mice.
British journal of pharmacology (2016-09-15)
Fiorentina Roviezzo, Vincenzo Brancaleone, Valentina Mattera Iacono, Antonio Bertolino, Giovanna De Cunto, Valentina Vellecco, Giuseppe Lungarella, Monica Lucattelli, Giuseppe Cirino
ABSTRACT
The majority of the severe vascular complications in fibrosis are a consequence of a deregulated activity of mediators controlling vasomotor tone. One of the most important of these mediators is endothelin-1 (ET-1). Here, we have investigated the role of proteinase-activated receptor 2 (PAR2) in the vascular dysfunction in a model of fibrosis, using tight-skin (Tsk) mice. Aortas were collected from Tsk, transgenic over-expressing PAR2 (TgPAR2), PAR2 deficient (PAR2 In aortas from Tsk mice, responses to phenylephrine were reduced, contractions to ET-1 were increased and vasorelaxation to PAR2-AP was enhanced. These alterations matched changes observed in whole vessel architecture such as vascular fibre re-organization, increased collagen deposition and enhanced α-smooth muscle actin expression. Expression of both ET Our data show that PAR2 counterbalanced enhanced contractions to ET-1 in aortas from Tsk mice. PAR2 could represent a possible target for novel drugs in the treatment of vascular complications in fibrosis. This article is part of a themed section on Targeting Inflammation to Reduce Cardiovascular Disease Risk. To view the other articles in this section visit http://onlinelibrary.wiley.com/doi/10.1111/bph.v174.22/issuetoc and http://onlinelibrary.wiley.com/doi/10.1111/bcp.v82.4/issuetoc.
Sigma-Aldrich
PAR-2 (1-6) amide (mouse, rat) trifluoroacetate salt, ≥97% (HPLC)
Sigma-Aldrich
Acetylcholine bromide, ≥99%