MilliporeSigma
  • Akt2 ablation prolongs life span and improves myocardial contractile function with adaptive cardiac remodeling: role of Sirt1-mediated autophagy regulation.
Akt2 ablation prolongs life span and improves myocardial contractile function with adaptive cardiac remodeling: role of Sirt1-mediated autophagy regulation.
Aging cell (2017-07-07)
Jun Ren, Lifang Yang, Li Zhu, Xihui Xu, Asli F Ceylan, Wei Guo, Jian Yang, Yingmei Zhang
ABSTRACT
Aging is accompanied with unfavorable geometric and functional changes in the heart involving dysregulation of Akt and autophagy. This study examined the impact of Akt2 ablation on life span and cardiac aging as well as the mechanisms involved with a focus on autophagy and mitochondrial integrity. Cardiac geometry, contractile, and intracellular Ca
Sigma-Aldrich
Benzenesulfonyl fluoride, 99%