MilliporeSigma
  • Dynamic properties of imidazolium orthoborate ionic liquids mixed with polyethylene glycol studied by NMR diffusometry and impedance spectroscopy.
Dynamic properties of imidazolium orthoborate ionic liquids mixed with polyethylene glycol studied by NMR diffusometry and impedance spectroscopy.
Magnetic resonance in chemistry : MRC (2017-07-29)
Andrei Filippov, Nail Azancheev, Amal Gibaydullin, Shubhankar Bhattacharyya, Oleg N Antzutkin, Faiz Ullah Shah
ABSTRACT
We used
Sigma-Aldrich
1-Bromodecane, 98%
Sigma-Aldrich
1-Bromoheptane, 99%