MilliporeSigma
  • Evaluation of putative precursors of key 'reductive' compounds in wines post-bottling.
Evaluation of putative precursors of key 'reductive' compounds in wines post-bottling.
Food chemistry (2017-12-31)
Marlize Z Bekker, Eric N Wilkes, Paul A Smith
ABSTRACT
Precursors to hydrogen sulfide (H
Sigma-Aldrich
Ethyl thioacetate, ≥98%
Sigma-Aldrich
Sodium thiomethoxide, 95%
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
Ethyl methyl sulfide, 96%