MilliporeSigma
  • Personalized siRNA-Nanoparticle Systemic Therapy using Metastatic Lymph Node Specimens Obtained with EBUS-TBNA in Lung Cancer.
Personalized siRNA-Nanoparticle Systemic Therapy using Metastatic Lymph Node Specimens Obtained with EBUS-TBNA in Lung Cancer.
Molecular cancer research : MCR (2017-10-11)
Tatsuya Kato, Daiyoon Lee, Huang Huang, William Cruz, Hideki Ujiie, Kosuke Fujino, Hironobu Wada, Priya Patel, Hsin-Pei Hu, Kentaro Hirohashi, Takahiro Nakajima, Masaaki Sato, Mitsuhito Kaji, Kichizo Kaga, Yoshiro Matsui, Juan Chen, Gang Zheng, Kazuhiro Yasufuku
ABSTRACT
Inhibiting specific gene expression with siRNA provides a new therapeutic strategy to tackle many diseases at the molecular level. Recent strategies called high-density lipoprotein (HDL)-mimicking peptide-phospholipid nanoscaffold (HPPS) nanoparticles have been used to induce siRNAs-targeted delivery to
Sigma-Aldrich
MISSION® esiRNA, targeting human KIF11