MilliporeSigma
  • Non-innocent probes in direct sonication: Metal assistance in oxidative radical CH functionalization.
Non-innocent probes in direct sonication: Metal assistance in oxidative radical CH functionalization.
Ultrasonics sonochemistry (2017-11-16)
Joachim Demaerel, Sebastian Govaerts, Rony Rajan Paul, Tom Van Gerven, Wim M De Borggraeve
ABSTRACT
Direct sonication by means of ultrasound horns constitutes a widely used technique in chemical process technology. However, the direct contact between the metal probe and the reaction mixture does not always leave the chemical system unaffected. In this report, we study the tert-butyl hydroperoxide-mediated trifluoromethylation of heterocyclic structures, and the influence of sonication thereon. Metal leaching is observed during the process and further examined, showing that several metals can interfere significantly with the chemical reaction under study. Notably, vanadium metal was found to increase the reaction rate exceptionally well, rendering it a useful additive for this type of reactions. Ultimately, some mechanistic considerations are offered, to provide more insight into the nature of the catalytic effect of leached metals.
Sigma-Aldrich
Sodium triflinate, ≥95.0% (T)