MilliporeSigma
  • Gas chromatographic and mass spectrometric studies on urinary organic acids in a patient with congenital lactic acidosis due to pyruvate decarboxylase deficiency.
Gas chromatographic and mass spectrometric studies on urinary organic acids in a patient with congenital lactic acidosis due to pyruvate decarboxylase deficiency.
Clinica chimica acta; international journal of clinical chemistry (1977-06-01)
R A Chalmers, A M Lawson, O Borud
PMID19171
ABSTRACT
Detailed studies, using gas chromatography and mass spectrometric methods, of the urinary organic acids excreted by a patient with proven pyruvate decarboxylase deficiency are reported. In addition to the greatly-increased levels of lactate and pyruvate, marked elevation in the levels of 2-oxoglutaric, malic, and isocitric acids were observed, with associated increases 2-hydroxyglutaric, fumaric, succinic, and glyceric acids, and reduced citric acid excretion. The levels of excretion during clinically static and acute periods are compared to those in a normal neonate and normal infants. The metabolites observed indicate a probable defect in the oxidation of pyruvate by pyruvate dehydrogenase and suggest the presence of secondary defects in the tricarboxylic acid cycle. Studies of this type may enable the relatively rapid identification of the probable underlying enzyme deficiency in cases of congenital lactic acidosis, prior to confirmatory enzyme studies.
Sigma-Aldrich
D-2-Hydroxyglutarate (D2HG) Assay Kit, sufficient for 200 fluorometric reactions