MilliporeSigma
  • An ultrasensitive electrochemiluminescent immunosensor based on graphene oxide coupled graphite-like carbon nitride and multiwalled carbon nanotubes-gold for the detection of diclofenac.
An ultrasensitive electrochemiluminescent immunosensor based on graphene oxide coupled graphite-like carbon nitride and multiwalled carbon nanotubes-gold for the detection of diclofenac.
Biosensors & bioelectronics (2017-11-03)
Liuyi Hu, Jing Zheng, Kang Zhao, Anping Deng, Jianguo Li
ABSTRACT
In this study, a novel competition-type electrochemiluminescent (ECL) immunosensor for detecting diclofenac (DCF) was fabricated with graphene oxide coupled graphite-like carbon nitride (GO-g-C
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide, ≥97.0% (T)
Supelco
5-Hydroxydiclofenac, analytical standard