MilliporeSigma
Novel pyrrolocycloalkylpyrazole analogues as CB
Chemical biology & drug design (2017-07-05)
Battistina Asproni, Ilaria Manca, Giansalvo Pinna, Elena Cichero, Paola Fossa, Gabriele Murineddu, Paolo Lazzari, Giovanni Loriga, Gérard A Pinna
ABSTRACT
Novel 1,4-dihydropyrazolo[3,4-a]pyrrolizine-, 4,5-dihydro-1H-pyrazolo[4,3-g]indolizine- and 1,4,5,6-tetrahydropyrazolo[3,4-c]pyrrolo[1,2-a]azepine-3-carboxamide-based compounds were designed and synthesized for cannabinoid CB
Sigma-Aldrich
1-Aminohomopiperidine, 95%