MilliporeSigma
  • Different protein kinase C isoenzymes mediate inhibition of cardiac rapidly activating delayed rectifier K
Different protein kinase C isoenzymes mediate inhibition of cardiac rapidly activating delayed rectifier K
British journal of pharmacology (2017-09-25)
Xueli Liu, Yuhong Wang, Hua Zhang, Li Shen, Yanfang Xu
ABSTRACT
Elevated angiotensin II (Ang II) and sympathetic activity contributes to a high risk of ventricular arrhythmias in heart disease. The rapidly activating delayed rectifier K The whole-cell patch-clamp technique was used to record I A broad spectrum PKC inhibitor Gö6983 (not inhibiting PKCε), a selective cPKC inhibitor Gö6976 and a PKCα-specific inhibitor peptide, blocked the inhibition of I Our results indicated that inhibition of I
Sigma-Aldrich
Chromanol 293B, ≥98% (HPLC), powder
Sigma-Aldrich
E-4031, ≥98% (HPLC), lyophilized powder