MilliporeSigma
  • Bifunctionalized ordered mesoporous organosilica synthesized in deep eutectic solvent for extraction of triazine herbicides from watermelon.
Bifunctionalized ordered mesoporous organosilica synthesized in deep eutectic solvent for extraction of triazine herbicides from watermelon.
Journal of chromatography. A (2017-11-15)
Zan Li, Jun Jia, Mingwei Wang, Hua Zhang, Hongyuan Yan, Fengxia Qiao
ABSTRACT
A C
Sigma-Aldrich
Indole-2-acetic acid