MilliporeSigma
  • Social modeling of eating mediated by mirror neuron activity: A causal model moderated by frontal asymmetry and BMI.
Social modeling of eating mediated by mirror neuron activity: A causal model moderated by frontal asymmetry and BMI.
Behavioural brain research (2017-10-17)
Laura McGeown, Ron Davis
ABSTRACT
The social modeling of eating effect refers to the consistently demonstrated phenomenon that individuals tend to match their quantity of food intake to their eating companion. The current study sought to explore whether activity within the mirror neuron system (MNS) mediates the social modeling of eating effect as a function of EEG frontal asymmetry and body mass index (BMI). Under the guise of rating empathy, 93 female undergraduates viewed a female video confederate "incidentally" consume either a low or high intake of chips while electroencephalogram (EEG) activity was recorded. Subsequent ad libitum chip consumption was quantified. A first- and second-stage dual moderation model revealed that frontal asymmetry and BMI moderated an indirect effect of model consumption on participants' food consumption as mediated by MNS activity at electrode site C3, a