MilliporeSigma
  • Discovery of a novel calcium-sensitive fluorescent probe for α-ketoglutarate.
Discovery of a novel calcium-sensitive fluorescent probe for α-ketoglutarate.
Acta pharmacologica Sinica (2017-07-18)
Lin-Lin Gan, Lin-Hai Chen, Fa-Jun Nan
ABSTRACT
α-Ketoglutarate (α-KG), a pivotal metabolite in energy metabolism, has been implicated in nonalcoholic fatty liver disease (NAFLD) and several cancers. It is recently proposed that plasma α-KG is a surrogate biomarker of NAFLD. Here, we report the development of a novel "turn-on" chemosensor for α-KG that contains a coumarin moiety as a fluorophore. Using benzothiazole-coumarin (BTC) as inspiration, we designed a probe for calcium ion recognition that possesses a unique fluorophore compared with previously reported probes for α-KG measurement. This chemosensor is based on the specific Schiff base reaction and the calcium ion recognition property of the widely used calcium indicator BTC. The probe was synthesized, and a series of parallel experiments were conducted to optimize the chemical recognition process. Compared to the initial weak fluorescence, a remarkable 7.6-fold enhancement in fluorescence intensity (I/I
Sigma-Aldrich
6-Hydroxycoumarin, 96%