MilliporeSigma
  • Paeoniflorin inhibits proliferation of endometrial cancer cells via activating MAPK and NF-κB signaling pathways.
Paeoniflorin inhibits proliferation of endometrial cancer cells via activating MAPK and NF-κB signaling pathways.
Experimental and therapeutic medicine (2017-12-30)
Jianxin Zhang, Fengchun Wang, Huali Wang, Yanna Wang, Yan Wu, Hui Xu, Chen Su
ABSTRACT
Paeoniflorin (PAE), a principal bioactive component of