MilliporeSigma
  • Synergistic effects of dendritic cell targeting and laser-microporation on enhancing epicutaneous skin vaccination efficacy.
Synergistic effects of dendritic cell targeting and laser-microporation on enhancing epicutaneous skin vaccination efficacy.
Journal of controlled release : official journal of the Controlled Release Society (2017-09-19)
Yoan Machado, Sanne Duinkerken, Veronika Hoepflinger, Melissa Mayr, Evgeniia Korotchenko, Almedina Kurtaj, Isabel Pablos, Markus Steiner, Angelika Stoecklinger, Joyce Lübbers, Maximillian Schmid, Uwe Ritter, Sandra Scheiblhofer, Michael Ablinger, Verena Wally, Sarah Hochmann, Anna M Raninger, Dirk Strunk, Yvette van Kooyk, Josef Thalhamer, Richard Weiss
ABSTRACT
Due to its unique immunological properties, the skin is an attractive target tissue for allergen-specific immunotherapy. In our current work, we combined a dendritic cell targeting approach with epicutaneous immunization using an ablative fractional laser to generate defined micropores in the upper layers of the skin. By coupling the major birch pollen allergen Bet v 1 to mannan from S. cerevisiae via mild periodate oxidation we generated hypoallergenic Bet-mannan neoglycoconjugates, which efficiently targeted CD14
Sigma-Aldrich
Aminoguanidine hydrochloride, ≥98%