MilliporeSigma
Myotube migration to cover and shape the testis of
Biology open (2017-11-11)
Silke Rothenbusch-Fender, Katharina Fritzen, Maik C Bischoff, Detlev Buttgereit, Susanne F Oenel, Renate Renkawitz-Pohl
ABSTRACT
During
Sigma-Aldrich
Phalloidin–Atto 565, suitable for fluorescence, ≥80.0% (HPLC)
Sigma-Aldrich
Shields and Sang M3 Insect Medium, With L-glutamine, without potassium bicarbonate, powder, suitable for insect cell culture