MilliporeSigma
  • Endothelial cell adhesion, signaling, and morphogenesis in fibroblast-derived matrix.
Endothelial cell adhesion, signaling, and morphogenesis in fibroblast-derived matrix.
Matrix biology : journal of the International Society for Matrix Biology (2009-04-21)
Patricia A Soucy, Lewis H Romer
ABSTRACT
Extracellular matrix plays a critical role in cellular development by providing signaling cues that direct morphogenesis. In order to study both the cues that natural matrix provides and endothelial cell responses to that information, human fetal lung fibroblasts were used to produce a fibrous three-dimensional matrix. Following the removal of the fibroblasts by detergent extraction, protein and proteoglycan constituents of the remaining matrix were identified by immunofluorescence and immunoblotting. Matrix components included fibronectin, tenascin-C, collagen I, collagen IV, collagen VI, versican, and decorin. Colocalization analysis suggested that fibronectin was a uniquely distributed matrix protein. Morphology, three-dimensional matrix adhesions, and integrin-mediated signaling during vasculogenesis were then studied in human endothelial cells seeded onto the fibroblast-derived matrix. Elongated morphology and decreased cell area were noted, as compared with cells on fibronectin-coated coverslips. Cell-matrix adhesions contained vinculin, pY397-FAK, and pY410-p130Cas, and all of these colocalized more with fibronectin than tenascin-C, collagen I, or collagen VI. Additionally, the endothelial cells remodeled the fibroblast-derived matrix and formed networks of tubes with demonstrable lumens. Matrix adhesions in these tubes also predominantly colocalized with fibronectin. The pattern of membrane type 1 matrix metalloprotease expression in the endothelial cells suggested its involvement in the matrix remodeling that occurred during tubulogenesis. These results indicated that information in fibroblast-derived matrix promoted vasculogenic behavior.
Sigma-Aldrich
Fibronectin solution human fibroblasts, cell culture derived, ~0.5 mg/mL, sterile-filtered, BioReagent, suitable for cell culture