MilliporeSigma
  • Dehydrogenase-modified carbon-fiber microelectrodes for the measurement of neurotransmitter dynamics. 2. Covalent modification utilizing avidin-biotin technology.
Dehydrogenase-modified carbon-fiber microelectrodes for the measurement of neurotransmitter dynamics. 2. Covalent modification utilizing avidin-biotin technology.
Analytical chemistry (1993-03-01)
P Pantano, W G Kuhr
ABSTRACT
Dehydrogenase enzymes are immobilized onto carbon-fiber microelectrode surfaces via avidin-biotin technology and a covalently linked hydrophilic tether. The avidin-biotin coupling strategy allows the selectivity of the electrochemical measurement to be easily changed without reoptimizing the immobilization conditions. Optimized derivatization conditions are demonstrated for dimer, tetramer, and hexamer dehydrogenases. Nonelectroactive substrates (ethanol, glucose-6-phosphate, and glutamate) are quantitated through the detection of enzyme-generated NADH by fast scan cyclic staircase voltammetry (100 V/s). The glutamate dehydrogenase-modified microelectrode possesses a 300-ms response time with a detection limit of 0.5 mM glutamate and a 1-60 mM linear concentration range.
Sigma-Aldrich
Biotinamidohexanoic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt, ≥90% (TLC), powder
Sigma-Aldrich
Biotin 3-sulfo-N-hydroxysuccinimide ester sodium salt, ≥90% (TLC), powder