MilliporeSigma
  • The Effects of Laser with Different Parameters and Crosshatching on Fibroblast Adhesion and Proliferation to Implant Surfaces.
The Effects of Laser with Different Parameters and Crosshatching on Fibroblast Adhesion and Proliferation to Implant Surfaces.
The International journal of oral & maxillofacial implants (2017-05-19)
Hakki Çelebi, Pembegül Uyar Arpacı, Ilhami Çelik, Serhan Akman
ABSTRACT
To compare the adhesion and proliferation effect of HGF-1 cells on pure titanium disks when the surfaces are unprocessed (machined surfaces after slicing) or modified with a laser. Twenty-eight titanium disks were divided into four groups. Three surface topographies were created using an erbium fiber laser: group 1 (unidirectional application [no crosshatch]), group 2 (crosshatching in two directions), and group 3 (crosshatching in three directions). The samples in group 4 were unprocessed and served as controls. The surface roughness of samples was investigated with scanning electron microscopy (SEM) and measured with a mechanical surface profilometer. HGF-1 cell line was used to analyze the adhesion of fibroblasts on the sample surfaces. The authors used SEM, XTT, and acridine orange tests to determine the adhesion of HGF-1 onto specimen surfaces and the cell morphology after incubation for 72 hours. XTT results and surface roughness values for all specimens were analyzed using the Kruskal-Wallis test. Ra values of group 1 to group 4 (control) were 1.13, 0.26, 0.38, and 0.19, respectively. Crosshatching decreased the surface roughness values compared with unidirectional application (P = .003). Cell morphology observed in SEM showed that the elongation direction of cells resulted in all directions related with additional filopodia extentions within the crosshatch groups. Therefore, the highest cell viability was also detected in the three-directional crosshatch group (group 3) (P = .000) in XTT assay. According to the acridine orange test, higher cell numbers were seen in group 3, similar to the XTT findings (P = .86). Crosshatching significantly increased the cell-covered implant surfaces compared with the unidirectional group. Decreasing the surface Ra values via crosshatching helped spread the fibroblast over the implants in any direction, thus increasing cell proliferation and adhesion.
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, sodium pyruvate, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
SAFC
Dulbecco′s Phosphate Buffered Saline, liquid, sterile-filtered, DPBS Modified, without calcium, without magnesium, suitable for cell culture