MilliporeSigma
  • Bid-Induced Release of AIF/EndoG from Mitochondria Causes Apoptosis of Macrophages during Infection with
Bid-Induced Release of AIF/EndoG from Mitochondria Causes Apoptosis of Macrophages during Infection with
Frontiers in cellular and infection microbiology (2017-12-01)
Wei-Lin Hu, Hai-Yan Dong, Yang Li, David M Ojcius, Shi-Jun Li, Jie Yan
ABSTRACT
Leptospirosis is a global zoonotic infectious disease caused by pathogenic
Sigma-Aldrich
Z-Ile-Glu(O-ME)-Thr-Asp(O-Me) fluoromethyl ketone, ≥90% (TLC), powder
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)