MilliporeSigma
  • Fluorophore labeling of a cell-penetrating peptide induces differential effects on its cellular distribution and affects cell viability.
Fluorophore labeling of a cell-penetrating peptide induces differential effects on its cellular distribution and affects cell viability.
Biochimica et biophysica acta (2017-09-19)
Ditlev Birch, Malene Vinther Christensen, Dan Staerk, Henrik Franzyk, Hanne Mørck Nielsen
ABSTRACT
Cell-penetrating peptides constitute efficient delivery vectors, and studies of their uptake and mechanism of translocation typically involve fluorophore-labeled conjugates. In the present study, the influence of a number of specific fluorophores on the physico-chemical properties and uptake-related characteristics of penetratin were studied. An array of seven fluorophores belonging to distinct structural classes was examined, and the impact of fluorophore labeling on intracellular distribution and cytotoxicity was correlated to the physico-chemical properties of the conjugates. Exposure of several mammalian cell types to fluorophore-penetratin conjugates revealed a strong structure-dependent reduction in viability (1.5- to 20-fold lower IC
Sigma-Aldrich
1-Hydroxy-7-azabenzotriazole solution, ~0.6 M in DMF
Sigma-Aldrich
7-Methoxycoumarin-4-acetic acid, 97%