MilliporeSigma
  • Oxytocin-receptor-expressing neurons in the parabrachial nucleus regulate fluid intake.
Oxytocin-receptor-expressing neurons in the parabrachial nucleus regulate fluid intake.
Nature neuroscience (2017-12-01)
Philip J Ryan, Silvano I Ross, Carlos A Campos, Victor A Derkach, Richard D Palmiter
ABSTRACT
Brain regions that regulate fluid satiation are not well characterized, yet are essential for understanding fluid homeostasis. We found that oxytocin-receptor-expressing neurons in the parabrachial nucleus of mice (Oxtr
Sigma-Aldrich
Polyethyleneglycol Bisphenol A Epichlorohydrin Copolymer, mol wt 15,000-20,000 Da
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
Triton X-114, laboratory grade
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Clozapine N-oxide