MilliporeSigma
  • Deletion of eIF2β lysine stretches creates a dominant negative that affects the translation and proliferation in human cell line: A tool for arresting the cell growth.
Deletion of eIF2β lysine stretches creates a dominant negative that affects the translation and proliferation in human cell line: A tool for arresting the cell growth.
Cancer biology & therapy (2017-07-12)
Gabrielle Dias Salton, Claudia Cilene Fernandes Correia Laurino, Nicolás Oliveira Mega, Andrés Delgado-Cañedo, Niclas Setterblad, Maryvonnick Carmagnat, Ricardo Machado Xavier, Elizabeth Cirne-Lima, Guido Lenz, João Antonio Pêgas Henriques, Jomar Pereira Laurino
ABSTRACT
Eukaryote initiation factor 2 subunit β (eIF2β) plays a crucial role in regulation protein synthesis, which mediates the interaction of eIF2 with mRNA. eIF2β contains evolutionarily conserved polylysine stretches in amino-terminal region and a zinc finger motif in the carboxy-terminus. The gene eIF2β was cloned under tetracycline transcription control and the polylysine stretches were deleted by site-directed mutagenesis (eIF2βΔ3K). The plasmid was transfected into HEK 293 TetR cells. These cells were analyzed for their proliferative and translation capacities as well as cell death rate. Experiments were performed using gene reporter assays, western blotting, flow cytometry, cell sorting, cell proliferation assays and confocal immunofluorescence. eIF2βΔ3K affected negatively the protein synthesis, cell proliferation and cell survival causing G2 cell cycle arrest and increased cell death, acting in a negative dominant manner against the native protein. Polylysine stretches are also essential for eIF2β translocated from the cytoplasm to the nucleus, accumulating in the nucleolus and eIF2βΔ3K did not make this translocation. eIF2β is involved in the protein synthesis process and should act in nuclear processes as well. eIF2βΔ3K reduces cell proliferation and causes cell death. Since translation control is essential for normal cell function and survival, the development of drugs or molecules that inhibit translation has become of great interest in the scenario of proliferative disorders. In conclusion, our results suggest the dominant negative eIF2βΔ3K as a therapeutic strategy for the treatment of proliferative disorders and that eIF2β polylysine stretch domains are promising targets for this.
Sigma-Aldrich
Anti-Mouse IgG (Fc specific) F(ab′)2 fragment−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.
Roche
Cell Proliferation Kit II (XTT), liquid, pkg of 1 kit, suitable for cell analysis, suitable for tissue culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule) F(ab′)2 fragment–Cy3 antibody produced in sheep, affinity isolated antibody, buffered aqueous solution
Pricing and availability is not currently available.